Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTGCAAACTGTGCAGGGTCTC[A/G]GCAGATGCCCTGAGAATCTTTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SETD4 - SET domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007259.2 | 1494 | Intron | NP_001007260.1 | |||
NM_001007261.2 | 1494 | Intron | NP_001007262.1 | |||
NM_001286752.1 | 1494 | Silent Mutation | GCC,GCT | A,A 405 | NP_001273681.1 | |
NM_017438.4 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | NP_059134.1 | |
XM_011529636.1 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_011527938.1 | |
XM_011529637.1 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_011527939.1 | |
XM_011529638.2 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_011527940.1 | |
XM_011529639.1 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_011527941.1 | |
XM_011529640.2 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_011527942.1 | |
XM_011529642.1 | 1494 | Silent Mutation | GCC,GCT | A,A 405 | XP_011527944.1 | |
XM_011529643.1 | 1494 | Silent Mutation | GCC,GCT | A,A 405 | XP_011527945.1 | |
XM_011529644.1 | 1494 | Silent Mutation | GCC,GCT | A,A 405 | XP_011527946.1 | |
XM_017028403.1 | 1494 | Silent Mutation | GCC,GCT | A,A 429 | XP_016883892.1 | |
XM_017028404.1 | 1494 | Silent Mutation | GCC,GCT | A,A 405 | XP_016883893.1 | |
XM_017028405.1 | 1494 | Intron | XP_016883894.1 |