Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAATATGGACCAGGTAGCCCCT[A/G]TGGCTAACAGTTACAGAGGGACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164740 | ||||||||||||||||||||
Literature Links: |
ETS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETS2 - ETS proto-oncogene 2, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256295.1 | 333 | Missense Mutation | ATG,GTG | M,V 155 | NP_001243224.1 | |
NM_005239.5 | 333 | Missense Mutation | ATG,GTG | M,V 15 | NP_005230.1 | |
XM_005260935.1 | 333 | Missense Mutation | ATG,GTG | M,V 15 | XP_005260992.1 | |
XM_017028290.1 | 333 | Missense Mutation | ATG,GTG | M,V 15 | XP_016883779.1 |