Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGCGACCCTCACATTCCTACA[A/G]TGCTCTGTGTGGCATCTTAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604736 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP25 - ubiquitin specific peptidase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283041.1 | Intron | NP_001269970.1 | ||||
NM_001283042.1 | Intron | NP_001269971.1 | ||||
NM_013396.4 | Intron | NP_037528.3 | ||||
XM_005260949.4 | Intron | XP_005261006.1 | ||||
XM_006723992.3 | Intron | XP_006724055.1 | ||||
XM_011529529.1 | Intron | XP_011527831.1 | ||||
XM_011529531.2 | Intron | XP_011527833.1 | ||||
XM_011529533.2 | Intron | XP_011527835.1 | ||||
XM_011529535.2 | Intron | XP_011527837.1 | ||||
XM_017028321.1 | Intron | XP_016883810.1 | ||||
XM_017028322.1 | Intron | XP_016883811.1 | ||||
XM_017028323.1 | Intron | XP_016883812.1 | ||||
XM_017028324.1 | Intron | XP_016883813.1 | ||||
XM_017028325.1 | Intron | XP_016883814.1 | ||||
XM_017028327.1 | Intron | XP_016883816.1 | ||||
XM_017028328.1 | Intron | XP_016883817.1 | ||||
XM_017028329.1 | Intron | XP_016883818.1 |