Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGGTTAGAAAATATGTTTGTC[A/G]TGCAGTTCAGTTCCAGCTCCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGPAT3 - 1-acylglycerol-3-phosphate O-acyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037553.1 | Intron | NP_001032642.1 | ||||
NM_020132.4 | Intron | NP_064517.1 | ||||
XM_005261160.4 | Intron | XP_005261217.1 | ||||
XM_006724029.3 | Intron | XP_006724092.1 | ||||
XM_006724030.3 | Intron | XP_006724093.1 | ||||
XM_006724031.3 | Intron | XP_006724094.1 | ||||
XM_011529660.2 | Intron | XP_011527962.1 | ||||
XM_011529661.2 | Intron | XP_011527963.1 | ||||
XM_011529662.2 | Intron | XP_011527964.1 | ||||
XM_011529663.2 | Intron | XP_011527965.1 | ||||
XM_011529664.2 | Intron | XP_011527966.1 | ||||
XM_011529665.2 | Intron | XP_011527967.1 | ||||
XM_017028408.1 | Intron | XP_016883897.1 | ||||
XM_017028409.1 | Intron | XP_016883898.1 | ||||
XM_017028410.1 | Intron | XP_016883899.1 | ||||
XM_017028411.1 | Intron | XP_016883900.1 | ||||
XM_017028412.1 | Intron | XP_016883901.1 | ||||
XM_017028413.1 | Intron | XP_016883902.1 | ||||
XM_017028414.1 | Intron | XP_016883903.1 |