Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAAAACTGAAATTTTGTCCCAC[A/T]TAAGGGAGTTTCTTCTTCCCTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603076 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCG1 - ATP binding cassette subfamily G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004915.3 | 75 | Intron | NP_004906.3 | |||
NM_016818.2 | 75 | Intron | NP_058198.2 | |||
NM_207174.1 | 75 | Intron | NP_997057.1 | |||
NM_207627.1 | 75 | UTR 5 | NP_997510.1 | |||
NM_207628.1 | 75 | Intron | NP_997511.1 | |||
NM_207629.1 | 75 | Intron | NP_997512.1 | |||
XM_011529806.1 | 75 | Intron | XP_011528108.1 | |||
XM_011529807.2 | 75 | Intron | XP_011528109.1 |