Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTCGCGCAAGTGCGGCTCTGC[A/G]GGGAGGAGGGAGGGCCGAGGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609314 MIM: 608094 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101930094 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101930094 - uncharacterized LOC101930094 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPH1 - radial spoke head 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC37A1 - solute carrier family 37 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320537.1 | 29 | Intron | NP_001307466.1 | |||
NM_018964.3 | 29 | UTR 5 | NP_061837.3 | |||
XM_011529614.2 | 29 | Intron | XP_011527916.1 | |||
XM_011529615.1 | 29 | Intron | XP_011527917.1 | |||
XM_017028376.1 | 29 | Intron | XP_016883865.1 | |||
XM_017028377.1 | 29 | Intron | XP_016883866.1 | |||
XM_017028378.1 | 29 | Intron | XP_016883867.1 | |||
XM_017028379.1 | 29 | Intron | XP_016883868.1 | |||
XM_017028380.1 | 29 | Intron | XP_016883869.1 | |||
XM_017028381.1 | 29 | Intron | XP_016883870.1 | |||
XM_017028382.1 | 29 | Intron | XP_016883871.1 | |||
XM_017028383.1 | 29 | Intron | XP_016883872.1 |