Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATGTTGTGGTCACAGGAGTTT[A/C]CTGTTCACAGCAATGTGTGTGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2L13 - BCL2 like 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BID - BH3 interacting domain death agonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001196.3 | 1859 | UTR 3 | NP_001187.1 | |||
NM_001244567.1 | 1859 | UTR 3 | NP_001231496.1 | |||
NM_001244569.1 | 1859 | UTR 3 | NP_001231498.1 | |||
NM_001244570.1 | 1859 | UTR 3 | NP_001231499.1 | |||
NM_001244572.1 | 1859 | UTR 3 | NP_001231501.1 | |||
NM_197966.2 | 1859 | UTR 3 | NP_932070.1 | |||
NM_197967.2 | 1859 | UTR 3 | NP_932071.1 | |||
XM_017028906.1 | 1859 | Intron | XP_016884395.1 |