Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGAAGAGAAATAAAGAGGAGA[C/G]AGTTAAAGAGACCAGAATAGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CECR1 - cat eye syndrome chromosome region, candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282225.1 | 2335 | UTR 3 | NP_001269154.1 | |||
NM_001282226.1 | 2335 | UTR 3 | NP_001269155.1 | |||
NM_001282227.1 | 2335 | UTR 3 | NP_001269156.1 | |||
NM_001282228.1 | 2335 | UTR 3 | NP_001269157.1 | |||
NM_001282229.1 | 2335 | UTR 3 | NP_001269158.1 | |||
NM_177405.2 | 2335 | UTR 3 | NP_803124.1 | |||
XM_006724080.3 | 2335 | UTR 3 | XP_006724143.1 | |||
XM_011546133.1 | 2335 | UTR 3 | XP_011544435.1 |