Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTATACTGGCAACAGAGCTGG[C/T]AAACAGGGCTCTGTTGACTCACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR14L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRR14L - proline rich 14 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173566.2 | 9401 | UTR 3 | NP_775837.2 | |||
XM_006724219.3 | 9401 | UTR 3 | XP_006724282.1 | |||
XM_006724220.3 | 9401 | UTR 3 | XP_006724283.1 | |||
XM_006724221.2 | 9401 | UTR 3 | XP_006724284.1 | |||
XM_011530080.2 | 9401 | UTR 3 | XP_011528382.1 | |||
XM_011530081.2 | 9401 | UTR 3 | XP_011528383.1 | |||
XM_017028736.1 | 9401 | UTR 3 | XP_016884225.1 | |||
XM_017028737.1 | 9401 | UTR 3 | XP_016884226.1 |