Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGAGGGGGGACCCCAGGTGACT[A/G]TCAGGACCAGGGAAGTAATTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600863 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSNK1E - casein kinase 1 epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001894.4 | 2632 | UTR 3 | NP_001885.1 | |||
NM_152221.2 | 2632 | UTR 3 | NP_689407.1 |
LOC400927-CSNK1E - LOC400927-CSNK1E readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289912.1 | 2632 | UTR 3 | NP_001276841.1 |