Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAAAAACCAAGAAACAAAACGC[C/T]GTAGAGTTAATACTTCATTCCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIEF1 - mitochondrial elongation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304564.1 | Intron | NP_001291493.1 | ||||
NM_019008.5 | Intron | NP_061881.2 | ||||
XM_011530236.1 | Intron | XP_011528538.1 | ||||
XM_011530237.1 | Intron | XP_011528539.1 | ||||
XM_011530238.1 | Intron | XP_011528540.1 | ||||
XM_017028838.1 | Intron | XP_016884327.1 |