Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAAAGCTAGACATGAAACCTTT[G/T]GTGAGCATTTCCTGAATGCTTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608685 | ||||||||||||||||||||
Literature Links: |
RIBC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIBC2 - RIB43A domain with coiled-coils 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015653.4 | 232 | Missense Mutation | GCT,TCT | A,S 71 | NP_056468.3 | |
XM_005261524.4 | 232 | UTR 5 | XP_005261581.1 | |||
XM_011530126.2 | 232 | Intron | XP_011528428.1 | |||
XM_017028766.1 | 232 | Missense Mutation | GCT,TCT | A,S 71 | XP_016884255.1 |
SMC1B - structural maintenance of chromosomes 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291501.1 | 232 | Intron | NP_001278430.1 | |||
NM_148674.4 | 232 | Intron | NP_683515.4 | |||
XM_011530144.2 | 232 | Intron | XP_011528446.1 | |||
XM_011530145.2 | 232 | Intron | XP_011528447.1 |