Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTCGCGGATGCTCAAGTCGTAGA[C/T]GGCGCCGTCGAGCAGCACCGCGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602128 MIM: 602220 | ||||||||||||||||||||
Literature Links: |
GAS2L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAS2L1 - growth arrest specific 2 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASL10A - RAS like family 10 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006477.4 | 664 | Missense Mutation | ATC,GTC | I,V 52 | NP_006468.1 | |
XM_011529821.1 | 664 | Intron | XP_011528123.1 | |||
XM_011529822.1 | 664 | Intron | XP_011528124.1 | |||
XM_011529823.1 | 664 | Intron | XP_011528125.1 |