Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTCTGGGCTCCCTCATAAGCAG[C/T]CCCTGAGACAAATATTCAGTGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLR1 - toll like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003263.3 | 4005 | Intron | NP_003254.2 | |||
XM_005262662.4 | 4005 | UTR 3 | XP_005262719.1 | |||
XM_011513742.2 | 4005 | UTR 3 | XP_011512044.1 | |||
XM_011513743.2 | 4005 | UTR 3 | XP_011512045.1 | |||
XM_011513744.2 | 4005 | UTR 3 | XP_011512046.1 | |||
XM_011513745.2 | 4005 | UTR 3 | XP_011512047.1 | |||
XM_017008571.1 | 4005 | UTR 3 | XP_016864060.1 | |||
XM_017008572.1 | 4005 | UTR 3 | XP_016864061.1 |