Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTTTACAAAAATTGACACTA[C/T]TAAAACCAACATGATTATCTTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGDH - UDP-glucose 6-dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184700.1 | 2375 | UTR 3 | NP_001171629.1 | |||
NM_001184701.1 | 2375 | UTR 3 | NP_001171630.1 | |||
NM_003359.3 | 2375 | UTR 3 | NP_003350.1 | |||
XM_005262667.2 | 2375 | UTR 3 | XP_005262724.1 |