Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACCTATCAGATCTGTGCCTGGG[C/T]CCTACCTTAGGTAATACAGCCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR17 - WD repeat domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170710.4 | Intron | NP_733828.2 | ||||
NM_181265.3 | Intron | NP_851782.3 | ||||
XM_005262725.3 | Intron | XP_005262782.1 | ||||
XM_011531582.2 | Intron | XP_011529884.1 | ||||
XM_017007686.1 | Intron | XP_016863175.1 | ||||
XM_017007687.1 | Intron | XP_016863176.1 | ||||
XM_017007688.1 | Intron | XP_016863177.1 | ||||
XM_017007689.1 | Intron | XP_016863178.1 | ||||
XM_017007690.1 | Intron | XP_016863179.1 | ||||
XM_017007691.1 | Intron | XP_016863180.1 | ||||
XM_017007692.1 | Intron | XP_016863181.1 | ||||
XM_017007693.1 | Intron | XP_016863182.1 | ||||
XM_017007694.1 | Intron | XP_016863183.1 | ||||
XM_017007695.1 | Intron | XP_016863184.1 | ||||
XM_017007696.1 | Intron | XP_016863185.1 | ||||
XM_017007697.1 | Intron | XP_016863186.1 |