Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGTCGGGTCTTTAAAAATACA[A/G]GCCCCTAAAGCACTAAGGGCATGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612839 | ||||||||||||||||||||
Literature Links: |
TET2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TET2 - tet methylcytosine dioxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127208.2 | 76 | Intron | NP_001120680.1 | |||
NM_017628.4 | 76 | UTR 5 | NP_060098.3 | |||
XM_005263082.2 | 76 | Intron | XP_005263139.1 | |||
XM_006714242.3 | 76 | Intron | XP_006714305.1 | |||
XM_017008319.1 | 76 | Intron | XP_016863808.1 |