Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGTGGTTGGCATTGACCTCGG[A/C]TTTCTCAACTGCTACATTGCTGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610796 | ||||||||||||||||||||
Literature Links: |
HSPA4L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPA4L - heat shock protein family A (Hsp70) member 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317381.1 | 359 | Silent Mutation | GGA,GGC | G,G 40 | NP_001304310.1 | |
NM_001317382.1 | 359 | Intron | NP_001304311.1 | |||
NM_001317383.1 | 359 | Silent Mutation | GGA,GGC | G,G 9 | NP_001304312.1 | |
NM_014278.3 | 359 | Silent Mutation | GGA,GGC | G,G 9 | NP_055093.2 | |
XM_011531745.2 | 359 | Silent Mutation | GGA,GGC | G,G 9 | XP_011530047.1 |
SLC25A31 - solute carrier family 25 member 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |