Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGTTATTAATAGCCAGCCACA[G/T]AGCCCTTCAAAATGTGGCTCCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD1 - SMAD family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003688.1 | Intron | NP_001003688.1 | ||||
NM_005900.2 | Intron | NP_005891.1 | ||||
XM_005262992.3 | Intron | XP_005263049.1 | ||||
XM_006714217.2 | Intron | XP_006714280.1 | ||||
XM_011531961.1 | Intron | XP_011530263.1 | ||||
XM_011531962.1 | Intron | XP_011530264.1 | ||||
XM_011531963.1 | Intron | XP_011530265.1 | ||||
XM_011531964.1 | Intron | XP_011530266.1 |
SMAD1-AS2 - SMAD1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |