Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGAGATAAACGTAGGATGCTGC[C/T]GGATTGTCAGACTGAAGAATGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FHDC1 - FH2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033393.2 | Intron | NP_203751.2 | ||||
XM_005263319.3 | Intron | XP_005263376.1 | ||||
XM_006714393.3 | Intron | XP_006714456.1 | ||||
XM_011532388.1 | Intron | XP_011530690.1 | ||||
XM_011532389.1 | Intron | XP_011530691.1 | ||||
XM_017008787.1 | Intron | XP_016864276.1 |
LOC729870 - uncharacterized LOC729870 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |