Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTATGTGCATTAATGGGAAAA[A/C]TTTTTGGAGGAAGCTGTTACTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCY1A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCY1A3 - guanylate cyclase 1 soluble subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000856.5 | Intron | NP_000847.2 | ||||
NM_001130682.2 | Intron | NP_001124154.1 | ||||
NM_001130683.3 | Intron | NP_001124155.1 | ||||
NM_001130684.2 | Intron | NP_001124156.1 | ||||
NM_001130685.2 | Intron | NP_001124157.1 | ||||
NM_001130687.2 | Intron | NP_001124159.1 | ||||
NM_001256449.1 | Intron | NP_001243378.1 | ||||
XM_005262955.2 | Intron | XP_005263012.1 | ||||
XM_005262956.2 | Intron | XP_005263013.1 | ||||
XM_005262957.2 | Intron | XP_005263014.1 | ||||
XM_006714196.2 | Intron | XP_006714259.1 | ||||
XM_006714197.2 | Intron | XP_006714260.1 | ||||
XM_006714198.2 | Intron | XP_006714261.1 | ||||
XM_011531900.2 | Intron | XP_011530202.1 |