Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATCCCTGCCAGTTTCTTGGGAG[A/G]CCTAGTGATCAACACTAACCATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP221 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP221 - cilia and flagella associated protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271049.1 | 470 | Intron | NP_001257978.1 | |||
XM_006712353.3 | 470 | Intron | XP_006712416.1 | |||
XM_017003550.1 | 470 | Intron | XP_016859039.1 | |||
XM_017003551.1 | 470 | Intron | XP_016859040.1 | |||
XM_017003552.1 | 470 | Intron | XP_016859041.1 | |||
XM_017003553.1 | 470 | Intron | XP_016859042.1 | |||
XM_017003554.1 | 470 | Intron | XP_016859043.1 | |||
XM_017003555.1 | 470 | Intron | XP_016859044.1 | |||
XM_017003556.1 | 470 | Intron | XP_016859045.1 | |||
XM_017003557.1 | 470 | Intron | XP_016859046.1 | |||
XM_017003558.1 | 470 | Intron | XP_016859047.1 | |||
XM_017003559.1 | 470 | Intron | XP_016859048.1 |
TMEM177 - transmembrane protein 177 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105198.1 | 470 | Missense Mutation | GAC,GGC | D,G 113 | NP_001098668.1 | |
NM_001105199.1 | 470 | Missense Mutation | GAC,GGC | D,G 113 | NP_001098669.1 | |
NM_030577.2 | 470 | Missense Mutation | GAC,GGC | D,G 113 | NP_085054.2 |