Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTTTGCTGTGAATATAGTTCC[G/T]TTTGGAAAAAGTTTCATATCTATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602452 | ||||||||||||||||||||
Literature Links: |
BUB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BUB1 - BUB1 mitotic checkpoint serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278616.1 | 2926 | Missense Mutation | AAA,AAC | K,N 938 | NP_001265545.1 | |
NM_001278617.1 | 2926 | Missense Mutation | AAA,AAC | K,N 901 | NP_001265546.1 | |
NM_004336.4 | 2926 | Missense Mutation | AAA,AAC | K,N 958 | NP_004327.1 |