Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTTTTCCATTTGGAAAGACAC[C/T]GTGGTAGGAAGGTAATTTAAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZEB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZEB2 - zinc finger E-box binding homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171653.1 | Intron | NP_001165124.1 | ||||
NM_014795.3 | Intron | NP_055610.1 | ||||
XM_006712881.3 | Intron | XP_006712944.1 | ||||
XM_006712882.3 | Intron | XP_006712945.1 | ||||
XM_011512232.2 | Intron | XP_011510534.1 | ||||
XM_017005414.1 | Intron | XP_016860903.1 | ||||
XM_017005415.1 | Intron | XP_016860904.1 |