Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAGCTCCAATGACTCACCAGGAG[A/G]ACCCAGGCCAAGGGAAGACTTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604462 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD39 - ankyrin repeat domain 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4C - semaphorin 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017789.4 | 3063 | UTR 3 | NP_060259.4 | |||
XM_006712606.3 | 3063 | Intron | XP_006712669.2 | |||
XM_011511378.2 | 3063 | UTR 3 | XP_011509680.1 | |||
XM_011511379.2 | 3063 | UTR 3 | XP_011509681.1 | |||
XM_011511380.1 | 3063 | UTR 3 | XP_011509682.1 | |||
XM_011511381.1 | 3063 | UTR 3 | XP_011509683.1 | |||
XM_011511382.2 | 3063 | UTR 3 | XP_011509684.1 | |||
XM_011511383.1 | 3063 | UTR 3 | XP_011509685.1 | |||
XM_017004393.1 | 3063 | UTR 3 | XP_016859882.1 | |||
XM_017004394.1 | 3063 | UTR 3 | XP_016859883.1 |