Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTTATTTTTGTTGGAGCTTTT[C/T]GCACTGCTGGGTTGTTTCTCCTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRXN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRXN1 - neurexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264642.3 | 5929 | Silent Mutation | GCA,GCG | A,A 1495 | XP_005264699.1 | |
XM_005264643.3 | 5929 | Silent Mutation | GCA,GCG | A,A 1480 | XP_005264700.1 | |
XM_006712137.3 | 5929 | Silent Mutation | GCA,GCG | A,A 1465 | XP_006712200.1 | |
XM_006712140.3 | 5929 | Intron | XP_006712203.2 | |||
XM_011533167.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1498 | XP_011531469.1 | |
XM_011533171.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1491 | XP_011531473.1 | |
XM_011533172.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1489 | XP_011531474.1 | |
XM_011533174.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1483 | XP_011531476.1 | |
XM_011533175.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1479 | XP_011531477.1 | |
XM_011533177.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1468 | XP_011531479.1 | |
XM_011533178.2 | 5929 | Silent Mutation | GCA,GCG | A,A 1468 | XP_011531480.1 | |
XM_011533180.2 | 5929 | Intron | XP_011531482.1 | |||
XM_011533183.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1209 | XP_011531485.1 | |
XM_017005303.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1507 | XP_016860792.1 | |
XM_017005304.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1506 | XP_016860793.1 | |
XM_017005305.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1504 | XP_016860794.1 | |
XM_017005306.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1503 | XP_016860795.1 | |
XM_017005307.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1501 | XP_016860796.1 | |
XM_017005308.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1500 | XP_016860797.1 | |
XM_017005309.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1498 | XP_016860798.1 | |
XM_017005310.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1497 | XP_016860799.1 | |
XM_017005311.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1492 | XP_016860800.1 | |
XM_017005312.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1490 | XP_016860801.1 | |
XM_017005313.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1488 | XP_016860802.1 | |
XM_017005314.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1487 | XP_016860803.1 | |
XM_017005315.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1486 | XP_016860804.1 | |
XM_017005316.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1486 | XP_016860805.1 | |
XM_017005317.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1484 | XP_016860806.1 | |
XM_017005318.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1483 | XP_016860807.1 | |
XM_017005319.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1482 | XP_016860808.1 | |
XM_017005320.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1479 | XP_016860809.1 | |
XM_017005321.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1477 | XP_016860810.1 | |
XM_017005322.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1474 | XP_016860811.1 | |
XM_017005323.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1469 | XP_016860812.1 | |
XM_017005324.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1459 | XP_016860813.1 | |
XM_017005325.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1456 | XP_016860814.1 | |
XM_017005326.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1455 | XP_016860815.1 | |
XM_017005327.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1450 | XP_016860816.1 | |
XM_017005328.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1444 | XP_016860817.1 | |
XM_017005329.1 | 5929 | Intron | XP_016860818.1 | |||
XM_017005330.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1436 | XP_016860819.1 | |
XM_017005331.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1431 | XP_016860820.1 | |
XM_017005332.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1424 | XP_016860821.1 | |
XM_017005333.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1421 | XP_016860822.1 | |
XM_017005334.1 | 5929 | Silent Mutation | GCA,GCG | A,A 1187 | XP_016860823.1 | |
XM_017005335.1 | 5929 | Silent Mutation | GCA,GCG | A,A 456 | XP_016860824.1 | |
XM_017005336.1 | 5929 | Silent Mutation | GCA,GCG | A,A 453 | XP_016860825.1 | |
XM_017005337.1 | 5929 | Silent Mutation | GCA,GCG | A,A 423 | XP_016860826.1 |