Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGTGCACAAGAATAAGACCTTA[A/T]GATAGGTAGGAGCAGGGTGAGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600034 MIM: 615572 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMX1 - empty spiracles homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFXN5 - sideroflexin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144579.2 | 3268 | UTR 3 | NP_653180.1 | |||
XM_005264648.1 | 3268 | UTR 3 | XP_005264705.1 | |||
XM_006712147.1 | 3268 | UTR 3 | XP_006712210.1 | |||
XM_011533186.2 | 3268 | UTR 3 | XP_011531488.1 | |||
XM_011533187.2 | 3268 | UTR 3 | XP_011531489.1 | |||
XM_011533188.2 | 3268 | UTR 3 | XP_011531490.1 | |||
XM_011533189.1 | 3268 | UTR 3 | XP_011531491.1 | |||
XM_011533190.2 | 3268 | UTR 3 | XP_011531492.1 | |||
XM_011533192.2 | 3268 | UTR 3 | XP_011531494.1 | |||
XM_011533194.2 | 3268 | UTR 3 | XP_011531496.1 | |||
XM_011533195.2 | 3268 | UTR 3 | XP_011531497.1 | |||
XM_011533196.2 | 3268 | UTR 3 | XP_011531498.1 | |||
XM_011533197.1 | 3268 | UTR 3 | XP_011531499.1 | |||
XM_017005340.1 | 3268 | UTR 3 | XP_016860829.1 | |||
XM_017005341.1 | 3268 | UTR 3 | XP_016860830.1 | |||
XM_017005342.1 | 3268 | UTR 3 | XP_016860831.1 | |||
XM_017005343.1 | 3268 | UTR 3 | XP_016860832.1 | |||
XM_017005344.1 | 3268 | UTR 3 | XP_016860833.1 | |||
XM_017005345.1 | 3268 | UTR 3 | XP_016860834.1 | |||
XM_017005346.1 | 3268 | UTR 3 | XP_016860835.1 |