Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTTAGCTCTATCTTAGCCTAGC[G/T]TTGTTAATGTTTGCAAATTGCTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600890 MIM: 143450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HADHA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HADHA - hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HADHB - hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000183.2 | Intron | NP_000174.1 | ||||
NM_001281512.1 | Intron | NP_001268441.1 | ||||
NM_001281513.1 | Intron | NP_001268442.1 | ||||
XM_011532803.1 | Intron | XP_011531105.1 | ||||
XM_017003939.1 | Intron | XP_016859428.1 |