Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATACTTATCTTATAGAAGTCAAA[G/T]AATGGAAGTACTTAGAACAATGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLIP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLIP4 - CAP-Gly domain containing linker protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287527.1 | Intron | NP_001274456.1 | ||||
NM_001287528.1 | Intron | NP_001274457.1 | ||||
NM_024692.5 | Intron | NP_078968.3 | ||||
XM_005264562.2 | Intron | XP_005264619.1 | ||||
XM_005264563.1 | Intron | XP_005264620.1 | ||||
XM_006712104.2 | Intron | XP_006712167.1 | ||||
XM_006712105.3 | Intron | XP_006712168.1 | ||||
XM_011533109.1 | Intron | XP_011531411.1 | ||||
XM_011533110.1 | Intron | XP_011531412.1 | ||||
XM_017004958.1 | Intron | XP_016860447.1 | ||||
XM_017004959.1 | Intron | XP_016860448.1 |