Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTACAGAGATCTGTTGGTAAT[C/G]TGAGGATTTTTATTCTACGTCGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609726 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YPEL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YPEL5 - yippee like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127399.1 | 511 | UTR 5 | NP_001120871.1 | |||
NM_001127400.1 | 511 | UTR 5 | NP_001120872.1 | |||
NM_001127401.1 | 511 | UTR 5 | NP_001120873.1 | |||
NM_016061.2 | 511 | Intron | NP_057145.1 | |||
XM_017004318.1 | 511 | UTR 5 | XP_016859807.1 | |||
XM_017004319.1 | 511 | Intron | XP_016859808.1 | |||
XM_017004320.1 | 511 | UTR 5 | XP_016859809.1 | |||
XM_017004321.1 | 511 | UTR 5 | XP_016859810.1 | |||
XM_017004322.1 | 511 | Intron | XP_016859811.1 |