Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGAATTCCTAAGCAGCCATGTT[C/G]TCACCATCACCGACCATGGCGCCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611458 | |||||||||||||||||||||||
Literature Links: |
GLB1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GLB1 - galactosidase beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000404.3 | 2263 | Intron | NP_000395.2 | |||
NM_001079811.2 | 2263 | Intron | NP_001073279.1 | |||
NM_001135602.2 | 2263 | Intron | NP_001129074.1 | |||
NM_001317040.1 | 2263 | Intron | NP_001303969.1 |
TMPPE - transmembrane protein with metallophosphoesterase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039770.2 | 2263 | UTR 3 | NP_001034859.2 | |||
NM_001136238.1 | 2263 | UTR 3 | NP_001129710.1 |