Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTGTTGAAGCTGCCTTAAATAC[C/T]GCAGAGGCAGACTGAAAAGCCCCTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
MIR8064 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MIR8064 - microRNA 8064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUSTN1 - musculoskeletal, embryonic nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM110 - transmembrane protein 110 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198563.2 | Intron | NP_940965.1 |
TMEM110-MUSTN1 - TMEM110-MUSTN1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198974.2 | Intron | NP_001185903.2 |