Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTTCAATAAAGACAGAAATAA[C/T]ATTTTCTCTCTTCTTCAGGGAGCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606807 | |||||||||||||||||||||||
Literature Links: |
IL17RD PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IL17RD - interleukin 17 receptor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318864.1 | 5409 | UTR 3 | NP_001305793.1 | |||
NM_017563.4 | 5409 | UTR 3 | NP_060033.3 | |||
XM_005265238.4 | 5409 | UTR 3 | XP_005265295.1 | |||
XM_006713209.3 | 5409 | UTR 3 | XP_006713272.1 | |||
XM_011533849.2 | 5409 | UTR 3 | XP_011532151.2 |