Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAATGGCATCTATGTGTGTCCG[C/T]AACTTTTCCATAGTGTCAGGTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616519 | ||||||||||||||||||||
Literature Links: |
DENND6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DENND6A - DENN domain containing 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152678.2 | 1820 | Silent Mutation | TTA,TTG | L,L 583 | NP_689891.1 | |
XM_006713019.3 | 1820 | Silent Mutation | TTA,TTG | L,L 582 | XP_006713082.1 | |
XM_006713020.2 | 1820 | Silent Mutation | TTA,TTG | L,L 517 | XP_006713083.1 | |
XM_017005864.1 | 1820 | Silent Mutation | TTA,TTG | L,L 516 | XP_016861353.1 |
PDE12 - phosphodiesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322176.1 | 1820 | Intron | NP_001309105.1 | |||
NM_001322177.1 | 1820 | Intron | NP_001309106.1 | |||
NM_177966.6 | 1820 | Intron | NP_808881.3 |