Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTCAGGAGGTTGAGAATAAGAA[A/G]TTCTAACATGTATACAGTATTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA18 - spermatogenesis associated 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297608.1 | Intron | NP_001284537.1 | ||||
NM_145263.3 | Intron | NP_660306.1 | ||||
XM_005265716.1 | Intron | XP_005265773.1 | ||||
XM_005265717.2 | Intron | XP_005265774.1 | ||||
XM_005265718.1 | Intron | XP_005265775.1 | ||||
XM_011534369.1 | Intron | XP_011532671.1 | ||||
XM_011534370.1 | Intron | XP_011532672.1 | ||||
XM_011534372.1 | Intron | XP_011532674.1 | ||||
XM_017007731.1 | Intron | XP_016863220.1 |