Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAGGATCTGGCTTGGAGTGCC[C/T]GTGTTAAGTGAACAGAGACTTGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
THG1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THG1L - tRNA-histidine guanylyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317824.1 | 62 | Intron | NP_001304753.1 | |||
NM_001317825.1 | 62 | Intron | NP_001304754.1 | |||
NM_001317826.1 | 62 | UTR 5 | NP_001304755.1 | |||
NM_017872.4 | 62 | Intron | NP_060342.2 | |||
XM_017009603.1 | 62 | Intron | XP_016865092.1 |