Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCAGGGCTTAAGCTCTTGTCTC[A/G]CTGCTGCTCCTACCATGTACAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1191 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1191 - KIAA1191 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079684.2 | Intron | NP_001073152.1 | ||||
NM_001079685.2 | Intron | NP_001073153.1 | ||||
NM_001287335.1 | Intron | NP_001274264.1 | ||||
NM_001287336.1 | Intron | NP_001274265.1 | ||||
NM_020444.4 | Intron | NP_065177.2 | ||||
XM_005265941.1 | Intron | XP_005265998.1 | ||||
XM_005265945.1 | Intron | XP_005266002.1 | ||||
XM_005265946.4 | Intron | XP_005266003.1 | ||||
XM_011534595.1 | Intron | XP_011532897.1 | ||||
XM_011534596.1 | Intron | XP_011532898.1 | ||||
XM_017009651.1 | Intron | XP_016865140.1 | ||||
XM_017009652.1 | Intron | XP_016865141.1 | ||||
XM_017009653.1 | Intron | XP_016865142.1 |
SIMC1 - SUMO interacting motifs containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |