Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCTCGAGTTTCATACATGCTA[C/T]TGTAAGGCCGCTTCCTCTCATCAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605974 MIM: 615253 | ||||||||||||||||||||
Literature Links: |
SLU7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLU7 - SLU7 homolog, splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006425.4 | 2160 | Missense Mutation | AAT,AGT | N,S 553 | NP_006416.3 | |
XM_005265793.3 | 2160 | Missense Mutation | AAT,AGT | N,S 563 | XP_005265850.1 | |
XM_011534418.2 | 2160 | Missense Mutation | AAT,AGT | N,S 553 | XP_011532720.1 |
ZBED8 - zinc finger BED-type containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |