Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCGGGACCCCTGCCAGCTGCAG[C/G]CGGGTGCCGTCGGCCAGCATGTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EXD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXD3 - exonuclease 3'-5' domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286823.1 | 3235 | Intron | NP_001273752.1 | |||
NM_017820.4 | 3235 | Silent Mutation | CGC,CGG | R,R 810 | NP_060290.3 | |
XM_011518810.1 | 3235 | Silent Mutation | CGC,CGG | R,R 965 | XP_011517112.1 | |
XM_011518814.1 | 3235 | Silent Mutation | CGC,CGG | R,R 521 | XP_011517116.1 | |
XM_011518815.1 | 3235 | Silent Mutation | CGC,CGG | R,R 521 | XP_011517117.1 | |
XM_017014861.1 | 3235 | Silent Mutation | CGC,CGG | R,R 1014 | XP_016870350.1 | |
XM_017014862.1 | 3235 | Silent Mutation | CGC,CGG | R,R 928 | XP_016870351.1 | |
XM_017014863.1 | 3235 | Silent Mutation | CGC,CGG | R,R 914 | XP_016870352.1 | |
XM_017014864.1 | 3235 | Silent Mutation | CGC,CGG | R,R 904 | XP_016870353.1 | |
XM_017014865.1 | 3235 | Silent Mutation | CGC,CGG | R,R 904 | XP_016870354.1 | |
XM_017014866.1 | 3235 | Silent Mutation | CGC,CGG | R,R 793 | XP_016870355.1 | |
XM_017014867.1 | 3235 | Silent Mutation | CGC,CGG | R,R 521 | XP_016870356.1 |
NRARP - NOTCH-regulated ankyrin repeat protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |