Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGAGGACTGGGCGCTGTGTGC[C/T]CTATTACCAGGGGCCCTCCAAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600844 MIM: 174762 | ||||||||||||||||||||
Literature Links: |
LRCOL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRCOL1 - leucine rich colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RX2 - purinergic receptor P2X 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282164.1 | 519 | Intron | NP_001269093.1 | |||
NM_001282165.1 | 519 | Missense Mutation | CCC,CTC | P,L 161 | NP_001269094.1 | |
NM_012226.4 | 519 | Missense Mutation | CCC,CTC | P,L 89 | NP_036358.2 | |
NM_016318.3 | 519 | Missense Mutation | CCC,CTC | P,L 137 | NP_057402.1 | |
NM_170682.3 | 519 | Missense Mutation | CCC,CTC | P,L 161 | NP_733782.1 | |
NM_170683.3 | 519 | Missense Mutation | CCC,CTC | P,L 161 | NP_733783.1 | |
NM_174872.2 | 519 | Missense Mutation | CCC,CTC | P,L 69 | NP_777361.1 | |
NM_174873.2 | 519 | Missense Mutation | CCC,CTC | P,L 161 | NP_777362.1 | |
XM_005266154.3 | 519 | Missense Mutation | CCC,CTC | P,L 161 | XP_005266211.1 | |
XM_005266155.4 | 519 | Missense Mutation | CCC,CTC | P,L 161 | XP_005266212.1 | |
XM_005266156.4 | 519 | Missense Mutation | CCC,CTC | P,L 161 | XP_005266213.1 | |
XM_011534786.2 | 519 | Missense Mutation | CCC,CTC | P,L 137 | XP_011533088.1 | |
XM_017019035.1 | 519 | Missense Mutation | CCC,CTC | P,L 161 | XP_016874524.1 |
POLE - DNA polymerase epsilon, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |