Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCAGGAATGTTCTGCCGATAC[C/T]GATAGGTCAGCTCACGGAAGCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHFR - checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161344.1 | 1805 | Missense Mutation | CAG,CGG | Q,R 615 | NP_001154816.1 | |
NM_001161345.1 | 1805 | Missense Mutation | CAG,CGG | Q,R 614 | NP_001154817.1 | |
NM_001161346.1 | 1805 | Missense Mutation | CAG,CGG | Q,R 603 | NP_001154818.1 | |
NM_001161347.1 | 1805 | Missense Mutation | CAG,CGG | Q,R 523 | NP_001154819.1 | |
NM_018223.2 | 1805 | Missense Mutation | CAG,CGG | Q,R 574 | NP_060693.2 |