Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCATACTAATCTTCAGATTGA[A/G]GAGGGGTTTTGTCCATTTTCTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGB1 - high mobility group box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313892.1 | 3671 | UTR 3 | NP_001300821.1 | |||
NM_001313893.1 | 3671 | UTR 3 | NP_001300822.1 | |||
NM_002128.5 | 3671 | UTR 3 | NP_002119.1 |