Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTGACTTACAAGTGTCCACTAA[C/T]GTTGTTAACAGCACAATAGGTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603581 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDH9 - protocadherin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318372.1 | 4894 | UTR 3 | NP_001305301.1 | |||
NM_001318373.1 | 4894 | UTR 3 | NP_001305302.1 | |||
NM_001318374.1 | 4894 | Intron | NP_001305303.1 | |||
NM_020403.4 | 4894 | UTR 3 | NP_065136.1 | |||
NM_203487.2 | 4894 | UTR 3 | NP_982354.1 | |||
XM_005266408.3 | 4894 | Intron | XP_005266465.1 | |||
XM_011535099.2 | 4894 | Intron | XP_011533401.1 | |||
XM_017020619.1 | 4894 | UTR 3 | XP_016876108.1 | |||
XM_017020620.1 | 4894 | Intron | XP_016876109.1 | |||
XM_017020621.1 | 4894 | Intron | XP_016876110.1 |