Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACAGGAATCGAATGGCAACGAC[A/G]GGTAGTAAGCACACAGCAACAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP8B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP8B1 - ATPase phospholipid transporting 8B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005603.4 | 3521 | Silent Mutation | CCC,CCT | P,P 1159 | NP_005594.1 | |
XM_006722481.3 | 3521 | Silent Mutation | CCC,CCT | P,P 1159 | XP_006722544.1 | |
XM_011526022.2 | 3521 | Silent Mutation | CCC,CCT | P,P 1159 | XP_011524324.1 | |
XM_011526023.2 | 3521 | Silent Mutation | CCC,CCT | P,P 1121 | XP_011524325.1 |
LOC100505549 - uncharacterized LOC100505549 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242804.1 | 3521 | Intron | NP_001229733.1 |