Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGGCGGCGGGGGCGGGTATGCC[A/G]GGCGGCGGCGGGGGGCCCTCGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZCCHC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC2 - zinc finger CCHC-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017742.5 | 23 | Silent Mutation | CCA,CCG | P,P 82 | NP_060212.4 | |
XM_006722493.3 | 23 | Silent Mutation | CCA,CCG | P,P 2 | XP_006722556.1 | |
XM_011526044.1 | 23 | Intron | XP_011524346.1 | |||
XM_017025802.1 | 23 | Silent Mutation | CCA,CCG | P,P 2 | XP_016881291.1 |