Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCATCCCCGAGACTCTCCGCACT[A/G]TCCTTACCCGTGACAGCCACTGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOBP - sine oculis binding protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018013.3 | 294 | UTR 5 | NP_060483.3 | |||
XM_005267041.4 | 294 | UTR 5 | XP_005267098.1 | |||
XM_005267042.4 | 294 | UTR 5 | XP_005267099.1 | |||
XM_011535920.2 | 294 | UTR 5 | XP_011534222.1 | |||
XM_011535921.2 | 294 | Intron | XP_011534223.1 | |||
XM_011535923.2 | 294 | Intron | XP_011534225.1 | |||
XM_017010991.1 | 294 | Intron | XP_016866480.1 | |||
XM_017010992.1 | 294 | Intron | XP_016866481.1 | |||
XM_017010993.1 | 294 | Intron | XP_016866482.1 | |||
XM_017010994.1 | 294 | Intron | XP_016866483.1 |