Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCTCTGTGGTGGAGGACAGCCT[C/T]GTTTTTCCCAGGCCCTCAGCTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WISP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WISP3 - WNT1 inducible signaling pathway protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003880.3 | 104 | Intron | NP_003871.1 | |||
NM_198239.1 | 104 | Intron | NP_937882.1 | |||
XM_011536220.1 | 104 | Intron | XP_011534522.1 | |||
XM_011536222.2 | 104 | UTR 5 | XP_011534524.2 | |||
XM_011536223.2 | 104 | Intron | XP_011534525.1 |