Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTTTTTACGTTGCTGCTCACTT[C/T]TCAGGGGGAAGGTTTGTTGGTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL10A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL10A1 - collagen type X alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000493.3 | 5743 | UTR 3 | NP_000484.2 | |||
XM_006715333.3 | 5743 | UTR 3 | XP_006715396.1 | |||
XM_011535432.2 | 5743 | UTR 3 | XP_011533734.1 | |||
XM_011535433.2 | 5743 | UTR 3 | XP_011533735.1 | |||
XM_017010248.1 | 5743 | UTR 3 | XP_016865737.1 | |||
XM_017010249.1 | 5743 | UTR 3 | XP_016865738.1 |
NT5DC1 - 5'-nucleotidase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152729.2 | 5743 | Intron | NP_689942.2 | |||
XM_006715378.3 | 5743 | Intron | XP_006715441.1 |