Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAAAACACCTATGGTGCAGTTT[G/T]TCAAGGCTGTTAGGTTAAGATTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THEMIS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THEMIS - thymocyte selection associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010923.2 | 3649 | UTR 3 | NP_001010923.1 | |||
NM_001164685.1 | 3649 | UTR 3 | NP_001158157.1 | |||
NM_001164687.1 | 3649 | UTR 3 | NP_001158159.1 | |||
NM_001318531.1 | 3649 | UTR 3 | NP_001305460.1 | |||
XM_011535813.2 | 3649 | UTR 3 | XP_011534115.1 | |||
XM_011535814.2 | 3649 | UTR 3 | XP_011534116.1 | |||
XM_011535816.1 | 3649 | UTR 3 | XP_011534118.1 | |||
XM_017010848.1 | 3649 | Intron | XP_016866337.1 | |||
XM_017010849.1 | 3649 | UTR 3 | XP_016866338.1 |