Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGAAGAGGTAACTAATCATTTT[A/G]TCAAGACGCAGAAAATAGATAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYB - MYB proto-oncogene, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130172.1 | Intron | NP_001123644.1 | ||||
NM_001130173.1 | Intron | NP_001123645.1 | ||||
NM_001161656.1 | Intron | NP_001155128.1 | ||||
NM_001161657.1 | Intron | NP_001155129.1 | ||||
NM_001161658.1 | Intron | NP_001155130.1 | ||||
NM_001161659.1 | Intron | NP_001155131.1 | ||||
NM_001161660.1 | Intron | NP_001155132.1 | ||||
NM_005375.3 | Intron | NP_005366.2 |